missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DC-SIGN+DC-SIGNR Antibody (120612) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB1621JF525
This item is not returnable.
View return policy
Description
DC-SIGN+DC-SIGNR Monoclonal antibody specifically detects DC-SIGN+DC-SIGNR in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,CyTOF
Specifications
| DC-SIGN+DC-SIGNR | |
| Monoclonal | |
| Janelia Fluor 525 | |
| CD209;CD209 antigen;CDSIGN;CLEC4L;DC-SIGN;DC-SIGN1;hDC-SIGN | |
| NIH-3T3 mouse embryonic fibroblast cell line transfected with human DC-SIGNR, Accession # Q9H2X3 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry, CyTOF | |
| 120612 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 30835 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction