Learn More
DC-SIGN/CD209 Antibody (MMD3), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB83451MFV500
394.40 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
Product Code. 18600170
  • £494.00 / 0.10mL
Description
DC-SIGN/CD209 Monoclonal antibody specifically detects DC-SIGN/CD209 in Mouse samples. It is validated for Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DC-SIGN/CD209
Monoclonal
mFluor Violet 500 SE
50mM Sodium Borate
Mouse
0.1 mL
Adaptive Immunity, Immunology, Virology Bacteria and Parasites
30835
Store at 4C in the dark.
Flow Cytometry, CyTOF
MMD3
Flow Cytometry, CyTOF-ready
CD209 antigendendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbingnon-integrin, CD209 molecule, CDSIGNHIV gpl20-binding protein, CLEC4LC-type lectin domain family 4 member L, DC-SIGN1C-type lectin domain family 4, member L, DC-SIGNMGC129965, Dendritic cell-specific ICAM-3-grabbing non-integrin 1
Purified mouse DC-SIGN/CD209 extracellular domain, Accession # FJ168685
RUO
Primary
Mouse
IgG2c