missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DC-SIGN/CD209 Antibody (MMD3), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB83451MFV450
394.40 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
DC-SIGN/CD209 Monoclonal antibody specifically detects DC-SIGN/CD209 in Mouse samples. It is validated for Flow Cytometry, CyTOF
Specifications
| DC-SIGN/CD209 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Adaptive Immunity, Immunology, Virology Bacteria and Parasites | |
| 30835 | |
| Store at 4C in the dark. |
| Flow Cytometry, CyTOF | |
| MMD3 | |
| Flow Cytometry, CyTOF-ready | |
| CD209 antigendendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbingnon-integrin, CD209 molecule, CDSIGNHIV gpl20-binding protein, CLEC4LC-type lectin domain family 4 member L, DC-SIGN1C-type lectin domain family 4, member L, DC-SIGNMGC129965, Dendritic cell-specific ICAM-3-grabbing non-integrin 1 | |
| Purified mouse DC-SIGN/CD209 extracellular domain, Accession # FJ168685 | |
| RUO | |
| Primary | |
| Mouse | |
| IgG2c |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction