missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ DC-SIGN/CD209 Antibody (INSERM patent anti-DC-SIGN) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28415-1mg
This item is not returnable.
View return policy
Description
DC-SIGN/CD209 Monoclonal antibody specifically detects DC-SIGN/CD209 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| DC-SIGN/CD209 | |
| Monoclonal | |
| Unconjugated | |
| Q9NNX6 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| INSERM patent anti-DC-SIGN | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| CD209 antigendendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbingnon-integrin, CD209 molecule, CDSIGNHIV gpl20-binding protein, CLEC4LC-type lectin domain family 4 member L, DC-SIGN1C-type lectin domain family 4, member L, DC-SIGNMGC129965, Dendritic cell-specific ICAM-3-grabbing non-integrin 1 | |
| DC-SIGN/ CD209 | |
| 1 mg | |
| Adaptive Immunity, Immunology, Virology Bacteria and Parasites | |
| 30835 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction