missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DC-LAMP Rabbit anti-Human, Clone: 7, Novus Biologicals™
Rabbit Monoclonal Antibody
£296.00 - £429.00
Specifications
| Antigen | DC-LAMP |
|---|---|
| Clone | 7 |
| Dilution | Immunocytochemistry/Immunofluorescence 1:20-1:100 |
| Applications | Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18241116
|
Novus Biologicals
NBP2-89553-50UL |
50 μg |
£296.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18264678
|
Novus Biologicals
NBP2-89553-100UL |
100 μg |
£429.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DC-LAMP Monoclonal specifically detects DC-LAMP in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| DC-LAMP | |
| Immunocytochemistry/Immunofluorescence 1:20-1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD208 antigen, DC-LAMP, DCLAMPDC-lysosome-associated membrane glycoprotein, LAMP, LAMP-3, lysosomal-associated membrane protein 3DC LAMP, lysosome-associated membrane glycoprotein 3, Protein TSC403, TSC403CD208 | |
| Recombinant Human DC-LAMP protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 7 | |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Dendritic Cell Markers | |
| 0.2 um filtered solution in PBS | |
| 27074 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title