Learn More
DC-LAMP Antibody (020), Allophycocyanin, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP2-89282APC
Product Code. 18678990
  • £418.00 / 0.10mL
Description
DC-LAMP Monoclonal antibody specifically detects DC-LAMP in Human samples. It is validated for ELISA, ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DC-LAMP
Monoclonal
APC
PBS
Rabbit
Protein A purified
RUO
Primary
Human
Purified
ELISA, Sandwich ELISA
020
ELISA, Sandwich ELISA Capture
CD208 antigen, DC-LAMP, DCLAMPDC-lysosome-associated membrane glycoprotein, LAMP, LAMP-3, lysosomal-associated membrane protein 3DC LAMP, lysosome-associated membrane glycoprotein 3, Protein TSC403, TSC403CD208
This antibody was obtained from a rabbit immunized with purified, recombinant Human DC-LAMP (Accession#: NP_055213.2; Lys28-Thr381).
0.1 mL
Dendritic Cell Markers
27074
Store at 4C in the dark.