missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DC-LAMP Antibody (007), Alexa Fluor™ 647, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89553AF647
This item is not returnable.
View return policy
Description
DC-LAMP Monoclonal specifically detects DC-LAMP in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| DC-LAMP | |
| Monoclonal | |
| Alexa Fluor 647 | |
| CD208 antigen, DC-LAMP, DCLAMPDC-lysosome-associated membrane glycoprotein, LAMP, LAMP-3, lysosomal-associated membrane protein 3DC LAMP, lysosome-associated membrane glycoprotein 3, Protein TSC403, TSC403CD208 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human DC-LAMP (NP_055213.2; Lys28-Thr381). | |
| 0.1 mL | |
| Dendritic Cell Markers | |
| 27074 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunofluorescence | |
| 7 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction