missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ DC-Cholesterol hydrochloride
GREENER_CHOICE

Product Code. 30000033
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 30000033

Brand: Tocris Bioscience™ 7176/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cationic cholesterol derivative

DC-Cholesterol hydrochloride is a cationic cholesterol derivative for use as a liposomal transfection reagent. Combines with the 'helper lipid' DOPE (Cat. No. 7175) after sonication or microfluidization to form unilamellar liposomes. Can be used in vitro and in vivo for transfer of nucleic acids, protein complexes, or other small molecule complexes into tissues, tumors and cells.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (3β)-3-[N-[2-(Dimethylamino)ethyl]carbamate cholest-5-en-3-ol hydrochloride
CAS 166023-21-8
Quantity 50 mg
Target Transfection Reagent
Molecular Formula C32H56N2O2.HCl
Purity 0.95
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.