Learn More
Specac™ DC-3 Diamond Compression Cell
Product Code. 15279937
Encompass_Preferred
Change view
Click to view available options
Diameter (Metric):
1.5 mm
Product Code. Diameter (Metric)
15279937 1.5 mm
1 options
Product Code. 15279937 Supplier Specac™ Supplier No. GS02555
Only null left
Add to basket
Add to basket
A Universal Diamond Compression Cell designed for FTIR microscopes. Enables samples to be compressed to an ideal thickness for transmission experiments. Uses 2 type IIIa diamond windows mounted onto a hastelloy plate. Clear aperture of 1.5mm diameter. High working pressure, high throughput.
Specifications
Diameter (Metric) 1.5 mm