missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ DC 271
GREENER_CHOICE

Product Code. 18717356
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 18717356

Brand: Tocris Bioscience™ 6873/5

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Fluorescent retinoic acid analog; solvochromatic probe

DC 271 is a fluorescent retinoic acid analog (retinoid all-trans-retinoic acid; ATRA) (Kd = 42 nM at cellular retinoid binding protein II (CRABPII)). Solvochromatic probe: fluorescent λex, λem, and quantum yield affected by solvent type. Activates same genes as endogenous retinoic acid in human keratinocyte cells (HaCaT) when incubated for 72 h at 1 μM. Also elicits cellular responses consistent with those of retinoic acid. Excitation λ = 351, 350, 382 and 378 nm in DMSO, EtOH, DCM and Toluene, respectively; emission λ = 442, 461, 572 and 447 nm in DMSO, EtOH, DCM and Toluene, respectively. Quantum yield = 19.3, 1.14, 71.45, 80.21 % in DMSO, EtOH, DCM and Toluene, respectively
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-[2-[1,2,3,4-Tetrahydro-4,4-dimethyl-1-(1-methylethyl)-6-quinolinyl]ethynyl]benzoic acid
CAS 198696-03-6
Quantity 5 mg
Target Fluorescent Receptor Probes
Molecular Formula C23H25NO2
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.