missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DBX2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564261
This item is not returnable.
View return policy
Description
Immunogen sequence: SSPRWRENSP EPSERLIQES SGAPPPEANS LQGALYLCSE EEAGSKGVLT G Highest antigen sequence identity to the following orthologs: Mouse - 49%, Rat - 53%.
DBX2 is a member of the developing brain homeobox (DBX) protein family, but while the related protein DBX1 is expressed in various regions of the developing brain, DBX2 shows a more restricted pattern of expression in the brain, and is also expressed in some mesenchymal cells such as limb buds and tooth germs. It is thought that DBX1 and DBX2 promote the development of a subset of interneurons, some of which help mediate left-right coordination of locomotor activity. In Xenopus, DBX2 is involved in primary neurogenesis and early neural plate patterning, and is thought to act as a cross-repressive partner of NKX6-2 in the patterning of the ventral neural tube.
Specifications
| DBX2 | |
| Polyclonal | |
| Unconjugated | |
| DBX2 | |
| 9430056A22Rik; 9630005K15; AI846217; DBX2; developing brain homeobox 2; developing brain homeobox protein 2; Gm1229; Homeobox protein DBX2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 440097 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6ZNG2 | |
| DBX2 | |
| Recombinant protein corresponding to Human DBX2. Recombinant protein control fragment (Product #RP-103622). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction