missing translation for 'onlineSavingsMsg'
Learn More

DBU, TRC

Product Code. 30008923
Change view
Click to view available options
Quantity:
10 g
50 g
150 g
Unit Size:
10g
150g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008923 50 g 50g
30004063 10 g 10g
30008093 150 g 150g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008923 Supplier Toronto Research Chemicals Supplier No. TRCD41709050G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6674-22-2
Formula Weight 152.1313
InChI Formula InChI=1S/C9H16N2/c1-2-5-9-10-6-4-8-11(9)7-3-1/h1-8H2
IUPAC Name 2,3,4,6,7,8,9,10-octahydropyrimido[1,2-a]azepine
Molecular Formula C9 H16 N2
Molecular Weight (g/mol) 152.24
Percent Purity >95%
SMILES C1CCN2CCCN=C2CC1
Synonym 1,8-Diazabicyclo[5.4.0]undec-7-ene, Pyrimido[1,2-a]azepine, 2,3,4,6,7,8,9,10-octahydro-, 2,3,4,6,7,8,9,10-Octahydropyrimido[1,2-a]azepine, 1,8-Diaza-7-bicyclo[5.4.0]undecene, 1,8-Diazabicyclo[5.4.0]undec-7-ene, 1,8-Diazabicyclo[5.4.0]undeca-7-ene, 1,8-Diazabicyclo[5.4.0]undecene-7, 1,8-Diazabicycloundec-7-ene, 2H,3H,4H,6H,7H,8H,9H,10H-Pyrimido[1,2-a]azepine, Active D 10, Alcanpoudre DBU 70-3KG, Amicure DBUE, DBU, Dabco DBU, Diazabicyclo[5.4.0]undec-7-ene, Lupragen N 700, NSC 111184, NSC 230466, PC CAT DBU, Polycat DBU, San-apro, U-CAT SA 851, DBU (GR53261X)
Chemical Name or Material 1,8-Diaza-7-bicyclo[5.4.0]undecene
Grade HPLC
Show More Show Less

Specifications

Quantity 50 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.