missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DBT Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£139.00 - £281.00
Specifications
| Antigen | DBT |
|---|---|
| Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18317882
|
Bio-Techne
NBP3-16010-20UL |
20 μg |
£139.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18342057
|
Novus Biologicals
NBP3-16010-100UL |
100 μg |
£281.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DBT Polyclonal antibody specifically detects DBT in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| DBT | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Stem Cell Markers | |
| PBS, 50% glycerol, pH7.3 | |
| 1629 | |
| IgG | |
| Affinity purified |
| Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| BCATE2BCKAD E2 subunit, BCKADE2, BCKAD-E2, branched chain acyltransferase, E2 component, Branched-chain alpha-keto acid dehydrogenase complex component E2, Dihydrolipoamide acetyltransferase component of branched-chain alpha-keto aciddehydrogenase complex, Dihydrolipoamide branched chain transacylase, dihydrolipoamide branched chain transacylase (E2 component of branched chainketo acid dehydrogenase complex; maple syrup urine disease), dihydrolipoamide branched chain transacylase E2, dihydrolipoyl transacylase, Dihydrolipoyllysine-residue (2-methylpropanoyl)transferase, E2, E2 component of branched chain alpha-keto acid dehydrogenase complex, E2B, EC 2.3.1.168, lipoamide acyltransferase component of branched-chain alpha-keto aciddehydrogenase complex, mitochondrial, lipoamide acyltransferase component of mitochondrial branched-chain alpha-ketoacid dehydrogenase complex, MGC9061, mitochondrial branched chain alpha-keto acid dehydrogenase transacylase subunit(E2b) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 11-247 of human DBT (NP_001909.3). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title