missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DBR1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16019-1-AP-150UL
This item is not returnable.
View return policy
Description
The RNA lariat debranching enzyme, or DBR1, specifically hydrolyzes 2-prime-to-5-prime branched phosphodiester bonds at the branch point of excised lariat intron RNA and converts them into linear molecules.Specifications
| DBR1 | |
| Polyclonal | |
| Unconjugated | |
| DBR1 | |
| DBR1, Lariat debranching enzyme | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 51163, 681234, 83703 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q923B1, Q9UK59 | |
| DBR1 | |
| DBR1 Fusion Protein Ag8830 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction