Learn More
DBR1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16019-1-AP-20UL
 View more versions of this product
Product Code. 16875464
  • £135.00 / 20µL
Description
The RNA lariat debranching enzyme, or DBR1, specifically hydrolyzes 2-prime-to-5-prime branched phosphodiester bonds at the branch point of excised lariat intron RNA and converts them into linear molecules.
Specifications
Specifications
DBR1
Polyclonal
Unconjugated
DBR1
DBR1, Lariat debranching enzyme
Rabbit
Antigen Affinity Chromatography
RUO
51163, 681234, 83703
-20°C
Liquid
Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry
0.37 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q923B1, Q9UK59
DBR1
DBR1 Fusion Protein Ag8830
20 μL
Primary
Mouse, Human, Rat