missing translation for 'onlineSavingsMsg'
Learn More

DBPR108, MedChemExpress

Product Code. 16487957
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16487957

Brand: Medchem Express HY12528

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress DBPR108 is a potent, selective, and orally bioavailable dipeptide-derived inhibitor of DPP4 with IC50 of 15 nM; no inhibition on DDP8 and DPP9.

DBPR108 is a potent, selective, and orally bioavailable dipeptide-derived inhibitor of DPP4 with IC50 of 15 nM

Specifications

Chemical Name or Material DBPR108
Physical Form Solid
CAS 1186426-66-3
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C16H25FN4O2
For Use With (Application) Metabolism-sugar/lipid metabolism
Solubility Information DMSO : ≥ 25 mg/mL (77.07 mM)
SMILES N#C[C@H]1N(C(CNC(C)(C)CC(N2CCCC2)=O)=O)C[C@@H](F)C1
Molecular Weight (g/mol) 324.39
Formula Weight 324.39
Percent Purity 98.75%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.