missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
DBPA Polyclonal antibody specifically detects DBPA in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | DBPA |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 550 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate |
| Gene Alias | cold shock domain protein A, Cold shock domain-containing protein A, cold-shock domain protein A, CSDA1, dbpA, DBPAcold-shock domain containing A1, DNA-binding protein A, Single-strand DNA-binding protein NF-GMB, ZO-1-associated nucleic acid-binding protein, ZONAB |
| Host Species | Rabbit |
| Immunogen | The immunogen for this product maps to a region between residue 250 and 300 of human ZO-1-associated nucleic acid-binding protein using the numbering given in entry NP_003642.3 (GeneID 8531). |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?