missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DBP Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540405
This item is not returnable.
View return policy
Description
Peptide sequence: SSPGHAPARA ALGTASGHRA GLTSRDTPSP VDPDTVEVLM TFEPDPADLA Sequence homology: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rat: 100%; Sheep: 100%.
D-site-binding protein (DBP) is a member of the PAR bZIP (proline and acidic amino acid-rich basic leucine zipper) transcription factor family.
Specifications
| DBP | |
| Polyclonal | |
| Unconjugated | |
| Dbp | |
| albumin D box-binding protein; albumin D-element binding protein; albumin D-element-binding protein; D site albumin promoter binding protein; d site albumin promoter-binding protein 1; D site of albumin promoter (albumin D-box) binding protein; D site of albumin promoter binding protein; D site-binding protein; DABP; D-box binding PAR bZIP transcription factor; DBP; DBP/GC; DBP02; DBP-maf; epididymis secretory protein Li 51; GC; Gc protein-derived macrophage activating factor; GC, vitamin D binding protein; gc-globulin; GcMAF; Gc-MAF; GRD3; group specific component; Group-specific component; group-specific component (vitamin D binding protein); HEL-S-51; taxREB302; Tax-responsive enhancer element-binding protein 302; Unknown (protein for MGC:129133); VDB; VDBG; VDBP; vitamin D-binding alpha-globulin; vitamin D-binding protein; Vitamin D-binding protein-macrophage activating factor | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 1628 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q10586 | |
| Dbp | |
| Synthetic peptide directed towards the middle region of human DBP. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction