missing translation for 'onlineSavingsMsg'
Learn More

dBET6, MedChemExpress

Product Code. 16453186
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16453186 25 mg 25mg
16450947 50 mg 50mg
16495486 100 mg 100mg
16434658 10 mg 10mg
16407377 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16453186 Supplier Medchem Express Supplier No. HY112588
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress dBET6 is a highly potent, selective and cell-permeable PROTAC connected by ligands for Cereblon and BET, with an IC50 of 14 nM, and has antitumor activity.

dBET6 is a highly potent, selective and cell-permeable PROTAC connected by ligands for Cereblon and BET, with an IC50 of 14 nM, and has antitumor activity.

Specifications

Chemical Name or Material dBET6
CAS 1950634-92-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C42H45ClN8O7S
Quantity 25 mg
Solubility Information DMSO : 50 mg/mL (59.43 mM; Need ultrasonic)
SMILES O=C(NCCCCCCCCNC(COC1=CC=CC(C(N2C(CC3)C(NC3=O)=O)=O)=C1C2=O)=O)C[C@H]4C5=NN=C(C)N5C6=C(C(C)=C(C)S6)C(C7=CC=C(Cl)C=C7)=N4
Molecular Weight (g/mol) 841.37
Formula Weight 841.37
Percent Purity 98.12%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.