missing translation for 'onlineSavingsMsg'
Learn More

dBET57, MedChemExpress

Product Code. 16461367
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16461367 50 mg 50mg
16427857 5 mg 5mg
16425138 10 mg 10mg
16405856 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16461367 Supplier Medchem Express Supplier No. HY123844
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress dBET57 is a potent and selective degrader of BRD4BD1 based on the PROTAC technology. dBET57 mediates recruitment to the CRL4Cereblon E3 ubiquitin ligase, with a DC50/5h of 500 nM for BRD4BD1, and is inactive on BRD4BD2.

dBET57 is a potent and selective degrader of BRD4BD1 based on the PROTAC technology. dBET57 mediates recruitment to the CRL4Cereblon E3 ubiquitin ligase, with a DC50/5h of 500 nM for BRD4BD1, and is inactive on BRD4BD2.

Specifications

Chemical Name or Material dBET57
CAS 1883863-52-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C34H31ClN8O5S
Quantity 50 mg
Solubility Information DMSO : 250 mg/mL (357.56 mM; Need ultrasonic)
SMILES O=C(NCCNC1=CC=CC(C(N2C(CC3)C(NC3=O)=O)=O)=C1C2=O)C[C@H]4C5=NN=C(C)N5C6=C(C(C)=C(C)S6)C(C7=CC=C(Cl)C=C7)=N4
Molecular Weight (g/mol) 699.18
Formula Weight 699.18
Percent Purity 99.66%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.