missing translation for 'onlineSavingsMsg'
Learn More

dBET1, MedChemExpress

Product Code. 16444696
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16444696

Brand: Medchem Express HY101838

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress dBET1 is a PROTAC connected by ligands for Cereblon and BRD4 with an EC50 of 430 nM. dBET1 is a PROTAC that composes of (+)-JQ1 (HY-13030) linked to NSC 527179 (HY-14658) with a linker.

dBET1 is a PROTAC connected by ligands for Cereblon and BRD4 with an EC50 of 430 nM. dBET1 is a PROTAC that composes of (+)-JQ1 (HY-13030) linked to NSC 527179 (HY-14658) with a linker.

Specifications

Chemical Name or Material dBET1
CAS 1799711-21-9
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C38H37ClN8O7S
Quantity 100 mg
Solubility Information DMSO : 43.33 mg/mL (55.18 mM; Need ultrasonic)
SMILES O=C(NCCCCNC(COC1=CC=CC(C(N2C(CC3)C(NC3=O)=O)=O)=C1C2=O)=O)C[C@H]4C5=NN=C(C)N5C6=C(C(C)=C(C)S6)C(C7=CC=C(Cl)C=C7)=N4
Molecular Weight (g/mol) 785.27
Formula Weight 785.27
Percent Purity 98.09%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.