missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DBCO-NHS Ester, TRC
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Specifications
Specifications
| Chemical Name or Material | DBCO-NHS Ester |
| CAS | 1353016-71-3 |
| Molecular Formula | C23H18N2O5 |
| Quantity | 1 mg |
| Synonym | 11,12-Didehydro-oxo-Dibenz[b,f]azocine-5(6H)-butanoic Acid 2,5-Dioxo-1-pyrrolidinyl Ester |
| InChI Formula | InChI=1S/C23H18N2O5/c26-20(13-14-23(29)30-25-21(27)11-12-22(25)28)24-15-18-7-2-1-5-16(18)9-10-17-6-3-4-8-19(17)24/h1-8H,11-15H2 |
| SMILES | O=C(CCC(=O)N1Cc2ccccc2C#Cc3ccccc13)ON4C(=O)CCC4=O |
| IUPAC Name | (2,5-dioxopyrrolidin-1-yl) 4-(2-azatricyclo[10.4.0.04,9]hexadeca-1(16),4,6,8,12,14-hexaen-10-yn-2-yl)-4-oxobutanoate |
| Molecular Weight (g/mol) | 402.4 |
| Formula Weight | 402.1216 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction