missing translation for 'onlineSavingsMsg'
Learn More

DBCO-NHS Ester, TRC

Product Code. 30366227
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 30366227

Brand: Toronto Research Chemicals TRCD19496550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DBCO-NHS Ester
CAS 1353016-71-3
Molecular Formula C23H18N2O5
Quantity 50 mg
Synonym 11,12-Didehydro-oxo-Dibenz[b,f]azocine-5(6H)-butanoic Acid 2,5-Dioxo-1-pyrrolidinyl Ester
InChI Formula InChI=1S/C23H18N2O5/c26-20(13-14-23(29)30-25-21(27)11-12-22(25)28)24-15-18-7-2-1-5-16(18)9-10-17-6-3-4-8-19(17)24/h1-8H,11-15H2
SMILES O=C(CCC(=O)N1Cc2ccccc2C#Cc3ccccc13)ON4C(=O)CCC4=O
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 4-(2-azatricyclo[10.4.0.04,9]hexadeca-1(16),4,6,8,12,14-hexaen-10-yn-2-yl)-4-oxobutanoate
Molecular Weight (g/mol) 402.4
Formula Weight 402.1216
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.