missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DBC1/p30 Antibody (PCRP-KIAA1967-1D10) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-24324JF669
This item is not returnable.
View return policy
Description
DBC1/p30 Monoclonal antibody specifically detects DBC1/p30 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry (Paraffin)
Specifications
| DBC1/p30 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| DBC.1, DBC1deleted in breast cancer 1, DBC-1p30 DBC protein, Deleted in breast cancer gene 1 protein, KIAA1967, NET35, p30 DBC, p30DBC | |
| Recombinant full-length human DBC1/p30 protein | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Tumor Suppressors | |
| 57805 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry (Paraffin) | |
| PCRP-KIAA1967-1D10 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction