missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DBC1/p30 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB200-189AF532
This item is not returnable.
View return policy
Description
DBC1/p30 Polyclonal antibody specifically detects DBC1/p30 in Human, Mouse samples. It is validated for Western Blot
Specifications
| DBC1/p30 | |
| Polyclonal | |
| Western Blot | |
| DBC.1, DBC1deleted in breast cancer 1, DBC-1p30 DBC protein, Deleted in breast cancer gene 1 protein, KIAA1967, NET35, p30 DBC, p30DBC | |
| A synthetic peptide, mapping to a region between residues 650 and 700 of human Deleted in Breast Cancer 1 using the numbering given in TrEMBL entry Q6P0Q9 (GeneID 57805). | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Tumor Suppressors | |
| 57805 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction