missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DBC1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92049-0.02ml
This item is not returnable.
View return policy
Description
DBC1 Polyclonal antibody specifically detects DBC1 in Human samples. It is validated for Western Blot
Specifications
| DBC1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| DBCCR1deleted in bladder cancer protein 1, deleted in bladder cancer 1, deleted in bladder cancer chromosome region candidate 1, FAM5AbA574M5.1 (deleted in bladder cancer chromosome region candidate 1 (IB3089A)), Protein FAM5A | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 20-300 of human BRINP1 (NP_055433.2). VQPSHQEPAGTDQHVSKEFDWLISDRGPFHHSRSYLSFVERHRQGFTTRYKIYREFARWKVRNTAIERRDLVRHPVPLMPEFQRSIRLLGRRPTTQQFIDTIIKKYGTHLLISATLGGEEALTMYMDKSRLDRKSGNATQSVEALHQLASSYFVDRDGTMRRLHEIQISTGAIKVTETRTGPLGCNSYDNLDSVSSVLLQSTESKLHLQGLQIIFPQYLQEKFVQSALSYIMCNGEGEYLCQNSQCRCQCAEEFPQCNCPITDIQIMEYTLANMAKSWAEA | |
| 0.02 mL | |
| Breast Cancer, Cancer, Cell Cycle and Replication, p53 Pathway, Tumor Suppressors | |
| 1620 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction