Learn More
Agilent Technologies DB-WAX, 20m x 0.1mm ID, 0.2um, FF Test
Brand:  Agilent Technologies 127-7023FF
Encompass_Preferred
DB-WAX, 20m x 0.1mm ID, 0.2um, FF Test
Product Code. 10498713
  • £872.10 / Each