missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DAZL Mouse anti-Human, Mouse, Rat, Cynomolgus Monkey, Clone: DAZL/4253, Novus Biologicals™
Mouse Monoclonal Antibody
£380.00
Specifications
| Antigen | DAZL |
|---|---|
| Clone | DAZL/4253 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1 to 2 μg/mL, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Description
DAZL Monoclonal specifically detects DAZL in Human, Mouse, Rat, Cynomolgus Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| DAZL | |
| 0.2 mg/mL | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat, Cynomolgus Monkey | |
| DAZHSPGY-like-autosomal, DAZL1DAZ-like autosomal, DAZLA, deleted in azoospermia-like, Deleted in azoospermia-like 1, germline specific RNA binding protein, MGC26406, spermatogenesis gene on the Y-like autosomal, SPGYLADAZ homolog | |
| Recognizes DAZL protein C terminus sequence CRVHHFRRSRAMLKSV. | |
| Primary | |
| Store at 4°C. |
| DAZL/4253 | |
| Western Blot 1 to 2 μg/mL, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 1618 | |
| IgG1 | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title