Learn More
DAZL Antibody (DAZL3/11A), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61937MFV500
This item is not returnable.
View return policy
Description
DAZL Monoclonal antibody specifically detects DAZL in Human, Mouse, Rat, Cynomolgus Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| Recognizes DAZL protein C terminus sequence CRVHHFRRSRAMLKSV | |
| Protein A purified | |
| Primary | |
| Human |