missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DAZL Antibody (DAZL3/11A), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61937AF488
This item is not returnable.
View return policy
Description
DAZL Monoclonal specifically detects DAZL in Human, Mouse, Rat, Cynomolgus Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| DAZL | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| DAZL | |
| Recognizes DAZL protein C terminus sequence CRVHHFRRSRAMLKSV | |
| 0.1 mL | |
| 1618 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
| DAZL3/11A | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| DAZHSPGY-like-autosomal, DAZL1DAZ-like autosomal, DAZLA, deleted in azoospermia-like, Deleted in azoospermia-like 1, germline specific RNA binding protein, MGC26406, spermatogenesis gene on the Y-like autosomal, SPGYLADAZ homolog | |
| Mouse | |
| Protein A purified | |
| Primary | |
| Cynomolgus, Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction