Learn More
DAZ2 Polyclonal Antibody
GREENER_CHOICE
DAZ2 Polyclonal Antibody
Brand:  Invitrogen PA5109617
Product Code. 17107553
  • £441.00 / 100µL
Description
DAZ2 Polyclonal Antibody for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DAZ2
Polyclonal
Unconjugated
DAZ2
DAZ2; deleted in azoospermia 2; deleted in azoospermia protein 2; pDP1678
Rabbit
Affinity Chromatography
RUO
57055
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
0.89 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q13117
DAZ2
Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human DAZ2 (NP_001005785.1).