missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
DAZ1 Polyclonal specifically detects DAZ1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | DAZ1 |
| Applications | Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | DyLight 594 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | deleted in azoospermia, deleted in azoospermia 1, FLJ17175, SPGYDAZdeleted in azoospermia protein 1 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human DAZ1. |
| Purification Method | Antigen Affinity-purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?