missing translation for 'onlineSavingsMsg'
Learn More

Emma Safety Footwear Daytona S1P Sandal

Product Code. 17572566
Change view
Click to view available options
Size:
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17572566 45 1 pair
17512566 39 1 pair
17522566 40 1 pair
17532566 41 1 pair
17542566 42 1 pair
17552566 43 1 pair
17562566 44 1 pair
17582566 46 1 pair
17592566 47 1 pair
17502576 48 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Product Code. 17572566 Supplier Emma Safety Footwear Supplier No. MM78754745
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Daytona is one of Emma's most breathable shoes. It's an ideal choice for warm, enclosed environments such as those in the Automotive industry.

  • Metal Free
  • Breathable and incredibly comfortable
  • It's not hard to see why the Daytona is one of Emma's most breathable shoes. It's an ideal choice for warm, enclosed environments such as those in the Automotive industry.
  • It features Emma's unique Hydro-Tec™ antibacterial technology, removing moisture and perspiration instantly.
  • Synthetic, reinforced toes and super swift velcro fasteners make the Daytona an ideal choice for all active occupations but especially for people working in Warehousing and Logistics.
  • Industries
    • Logistics & transport
    • Electronics
    • Manufacturing
  • Safety shoe
    • S1P-Safetyshoe with sole plate for dry work

Specifications

Certifications/Compliance ISO 20345:2011
Closure Type Velcro
Color Black, Gray
For Use With (Application) Logistics and Transport, Electronics, Manufacturing
Liner Material Hydro-Tec Antibacterial
Material Nubuck
Size 45
Fit Standard Width
Product Type Safety Shoe
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.