missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DAXX Recombinant Rabbit Monoclonal Antibody (23GB795), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | DAXX |
|---|---|
| Clone | 23GB795 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30087978
![]() |
Invitrogen™
MA554146 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087928
|
Invitrogen™
MA554147 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DAXX Recombinant Monoclonal antibody specifically detects DAXX in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| DAXX | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| O35613, Q8VIB2, Q9UER7 | |
| 13163, 140926, 1616 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB795 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| BING2; CENP-C binding protein; DADB-159G18.9; DAP6; Daxx; death domain associated protein; Death domain-associated protein 6; death-associated protein 6; death-domain associated protein; EAP1; ETS1-associated protein 1; Fas death domain-associated protein; Fas-binding protein; hDaxx; MGC126245; MGC126246; SMIM40; Unknown (protein for MGC:138055) | |
| A synthetic peptide of human DAXX (230-300AA). | |
| Primary | |
| -20°C | |
| DAXX |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title