missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Daxx Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77011-0.025mg
This item is not returnable.
View return policy
Description
Daxx Polyclonal specifically detects Daxx in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Daxx | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| BING2, CENP-C binding protein, DAP6MGC126245, Daxx, death domain-associated protein 6, death-associated protein 6, death-domain associated protein, EAP1, ETS1-associated protein 1, Fas death domain-associated protein, Fas-binding protein, hDaxx, MGC126246 | |
| Rabbit | |
| 120 kDa | |
| 0.025 mg | |
| Apoptosis, Cancer, Tumor Suppressors | |
| 1616 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 10 μg/mL | |
| CAG33366 | |
| DAXX | |
| Antibody was raised against a peptide corresponding to amino acids near the carboxy terminus of human Daxx. The immunogen is located within the last 50 amino acids of Daxx. Amino Acid Squence: TSVATQCDPEEIIVLSDSD | |
| Ion exchange chromatography | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction