missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Daxx Antibody (DAXX-01), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-491AF594
This item is not returnable.
View return policy
Description
Daxx Monoclonal specifically detects Daxx in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| Daxx | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| DAXX | |
| Recombinant C-terminal part (aa 558-740) of human Daxx. | |
| 0.1 mL | |
| Primary | |
| The antibody DAXX-01 reacts with Daxx, a death domain containing protein mainly expressed in fetal and adult human and mouse tissue. | |
| Store at 4°C in the dark. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| DAXX-01 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| BING2, CENP-C binding protein, DAP6MGC126245, Daxx, death domain-associated protein 6, death-associated protein 6, death-domain associated protein, EAP1, ETS1-associated protein 1, Fas death domain-associated protein, Fas-binding protein, hDaxx, MGC126246 | |
| Mouse | |
| Protein A purified | |
| Apoptosis, Cancer, Phospho Specific, Tumor Suppressors | |
| 1616.0 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction