missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DAX1/NR0B1 Antibody (OTI5F5), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70116B
This item is not returnable.
View return policy
Description
DAX1/NR0B1 Monoclonal specifically detects DAX1/NR0B1 in Human samples. It is validated for Immunohistochemistry.
Specifications
| DAX1/NR0B1 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| NR0B1 | |
| Recombinant protein fragment corresponding to amino acids 161-430 of human DAX1/NR0B1 (NP_000466) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Immunohistochemistry | |
| OTI5F5 | |
| Immunohistochemistry | |
| AHCDSS, AHCH, AHX, DAX1DAX-1, dosage-sensitive sex reversal, DSS-AHC critical region on the X chromosome protein 1, GTD, HHG, NROB1, nuclear hormone receptor, Nuclear receptor DAX-1, nuclear receptor subfamily 0 group B member 1, nuclear receptor subfamily 0, group B, member 1 | |
| Mouse | |
| Affinity Purified | |
| Cancer, GPCR | |
| 190.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction