missing translation for 'onlineSavingsMsg'
Learn More

DAX1/NR0B1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£292.00 - £488.00

Specifications

Antigen DAX1/NR0B1
Dilution Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL
Applications Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18691758
View Documents
Novus Biologicals
NBP2-68781-25ul
25 μL
£292.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18676308
View Documents
Novus Biologicals
NBP2-68781
100 μg
£488.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

DAX1/NR0B1 Polyclonal antibody specifically detects DAX1/NR0B1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications

Specifications

DAX1/NR0B1
Immunofluorescence
Unconjugated
Rabbit
Cancer
PBS (pH 7.2) and 40% Glycerol
190
IgG
Protein A purified
Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL
Polyclonal
Purified
RUO
Human
AHCDSS, AHCH, AHX, DAX1DAX-1, dosage-sensitive sex reversal, DSS-AHC critical region on the X chromosome protein 1, GTD, HHG, NROB1, nuclear hormone receptor, Nuclear receptor DAX-1, nuclear receptor subfamily 0 group B member 1, nuclear receptor subfamily 0, group B, member 1
This antibody was developed against a recombinant protein corresponding to amino acids: GKDHPRQGSILYSMLTSAKQTYAAPKAPEATLGPCWGCSCGSDPGVGRAGLPGGRPVALLYRCCF
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.