missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DATF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA536337
This item is not returnable.
View return policy
Description
This antibody detects endogenous protein at a molecular weight of 244 kDa. Purity is >95% by SDS-PAGE.
Dio-1 (Death Inducer-Obliterator-1) is a putative transcription factor that contains two zinc finger motifs. Dio-1 translocates to the nucleus, and activates apoptosis during limb development. Programmed cell death, a highly regulated form of apoptosis, plays an important role in determining the amount of tissue, the shape, and the definition of each digit during limb development. Dio-1 expression is upregulated when an apoptotic signal is detected, and subsequently apoptosis is induced. This process is similar to the expression of NFkB and NGF in response to external signals. Dio-1 expression is suppressed by caspase inhibitors and Bcl-2 expression. This supports the theory that Dio-1 functions in the onset of programmed cell death.
Specifications
| DATF1 | |
| Polyclonal | |
| Unconjugated | |
| DIDO1 | |
| 6720461J16Rik; BYE1; C20orf158; D130048F08Rik; DATF1; DATF-1; death associated transcription factor 1; death inducer-obliterator 1; death inducer-obliterator-2; death inducer-obliterator-3; death-associated transcription factor 1; Death-inducer obliterator 1; dido; Dido1; DIDO2; DIDO3; DIO1; DIO-1; dJ885L7.8; KIAA0333 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11083, 23856, 362286 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8C9B9, Q9BTC0 | |
| DIDO1 | |
| Synthetic peptide corresponding to amino acids 154-198 of human Dio-1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction