missing translation for 'onlineSavingsMsg'
Learn More

Datelliptium chloride, MedChemExpress

Product Code. p-200097214
Encompass_Preferred
Click to view available options
Quantity:
10 mg
20 mg
Unit Size:
10mg
20mg
This item is not returnable. View return policy

Product Code. 16440856

Brand: Medchem Express HYU00337

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Datelliptium chloride is a DNA-intercalating agent derived from ellipticine, with anti-tumor activities.

Datelliptium chloride is a DNA-intercalating agent derived from ellipticine, with anti-tumor activities.

Specifications

Chemical Name or Material Datelliptium chloride
CAS 105118-14-7
Color Red
Purity Grade Notes Research
Molecular Formula C23H28ClN3O
Quantity 20 mg
Solubility Information H2O : 41.67 mg/mL (104.71 mM; Need ultrasonic) ∣DMSO : 5 mg/mL (12.56 mM; Need ultrasonic)
SMILES CC1=C2C(NC3=C2C=C(O)C=C3)=C(C)C4=CC=[N+](CCN(CC)CC)C=C41.[Cl-]
Molecular Weight (g/mol) 397.94
Formula Weight 397.94
Percent Purity 98.33%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.