missing translation for 'onlineSavingsMsg'
Learn More

Datelliptium chloride, MedChemExpress

Product Code. 16440856
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
20 mg
Unit Size:
10mg
20mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16440856 20 mg 20mg
16410359 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16440856 Supplier Medchem Express Supplier No. HYU00337

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Datelliptium chloride is a DNA-intercalating agent derived from ellipticine, with anti-tumor activities.

Datelliptium chloride is a DNA-intercalating agent derived from ellipticine, with anti-tumor activities.

Specifications

Chemical Name or Material Datelliptium chloride
CAS 105118-14-7
Color Red
Purity Grade Notes Research
Molecular Formula C23H28ClN3O
Quantity 20 mg
Solubility Information H2O : 41.67 mg/mL (104.71 mM; Need ultrasonic) ∣DMSO : 5 mg/mL (12.56 mM; Need ultrasonic)
SMILES CC1=C2C(NC3=C2C=C(O)C=C3)=C(C)C4=CC=[N+](CCN(CC)CC)C=C41.[Cl-]
Molecular Weight (g/mol) 397.94
Formula Weight 397.94
Percent Purity 98.33%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.