missing translation for 'onlineSavingsMsg'
Learn More

Dasotraline hydrochloride, MedChemExpress

Product Code. 16465398
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16465398 10 mg 10mg
16461309 1 mg 1mg
16433336 25 mg 25mg
16418117 5 mg 5mg
16441587 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16465398 Supplier Medchem Express Supplier No. HY12850A
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Dasotraline hydrochloride (SEP-225289 hydrochloride) is a triple reuptake inhibitor that blocks dopamine, norepinephrine, and serotonin transporters with IC50 values of 4, 6, and 11 nM, respectively.

Dasotraline hydrochloride (SEP-225289 hydrochloride) is a triple reuptake inhibitor that blocks dopamine, norepinephrine, and serotonin transporters with IC50 values of 4, 6, and 11 nM, respectively.

Specifications

Chemical Name or Material Dasotraline hydrochloride
CAS 675126-08-6
Color White
Purity Grade Notes Research
Molecular Formula C16H16Cl3N
Quantity 10 mg
Synonym SEP-225289 hydrochloride
Solubility Information DMSO : ≥ 31 mg/mL (94.32 mM) ∣H2O : 1.61 mg/mL (4.90 mM; ultrasonic and warming and heat to 60°C)
SMILES N[C@@H]1CC[C@@H](C2=CC=C(Cl)C(Cl)=C2)C3=CC=CC=C31.Cl[H]
Molecular Weight (g/mol) 328.66
Formula Weight 328.66
Percent Purity 99.74%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.