missing translation for 'onlineSavingsMsg'
Learn More

Dasatinib hydrochloride, MedChemExpress

Product Code. 16490936
Encompass_Preferred
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
This item is not returnable. View return policy

Product Code. 16490936

Brand: Medchem Express HY10181A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Dasatinib (BMS-354825) hydrochloride is a highly potent, ATP competitive, orally active dual Src/Bcr-Abl inhibitor with potent antitumor activity. The Kis are 16 pM and 30 pM for Src and Bcr-Abl, respectively. Dasatinib hydrochloride inhibits Bcr-Abl and Src with IC50s of apoptosis and autophagy.

Dasatinib (BMS-354825) hydrochloride is a highly potent, ATP competitive, orally active dual Src/Bcr-Abl inhibitor with potent antitumor activity. The Kis are 16 pM and 30 pM for Src and Bcr-Abl, respectively. Dasatinib hydrochloride inhibits Bcr-Abl and Src with IC50s of apoptosis and autophagy.

Specifications

Chemical Name or Material Dasatinib hydrochloride
Physical Form Solid
CAS 854001-07-3
Color White
Quantity 200 mg
Purity Grade Notes Research
Molecular Formula C22H27Cl2N7O2S
For Use With (Application) Cancer-Kinase/protease
Synonym BMS-354825 hydrochloride
Solubility Information DMSO : 15 mg/mL (28.60 mM; Need ultrasonic and warming) ∣H2O : 10 mg/mL (19.07 mM; Need ultrasonic)
SMILES O=C(C1=CN=C(S1)NC2=NC(C)=NC(N3CCN(CC3)CCO)=C2)NC4=C(C=CC=C4Cl)C.[H]Cl
Molecular Weight (g/mol) 524.47
Formula Weight 524.47
Percent Purity 98.01%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.