missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Dasatinib
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
100 mg
missing translation for 'unitSize'
100mg
10mg
Description
- Dasatinib is a potent, ATP-competitive tyrosine kinase inhibitor
- It is specific for SRC/ABL kinases, for example, ABL, SRC, LCK, and YES with IC50 values of < 1.0, 0.5, 0.4 and 0.5 nM, respectively, and also demonstrates activity against KIT with an IC50 = 5.0 nM (Lombardo et al.; Davis et al.) Dasatinib is a second-generation inhibitor of the oncogenic tyrosine kinase BCR-ABL with 325-fold more potency than imatinib, and is also able to inhibit imatinib-resistant BCR-ABL mutants (Tokarski et al.) It also inhibits a large number of other kinases (76 of 148 kinases tested) when screened at 10 μM (Carter et al.)
Specifications
Specifications
| CAS | 302962-49-8 |
| Synonym | BMS 354825; Sprycel |
| Quantity | 10 mg |
| Molecular Formula | C22H26ClN7O2S |
| Purity | ≥98% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction