missing translation for 'onlineSavingsMsg'
Learn More

Darunavir Ethanolate, MedChemExpress

Product Code. 16433307
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16433307

Brand: Medchem Express HY17041

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Darunavir ethanolate (TMC114 Ethanolate) is a potent HIV protease inhibitor used to treat and prevent HIV/AIDS. Darunavir has a Ki of 1 nM for wild type HIV-1 protease.

Darunavir ethanolate (TMC114 Ethanolate) is a potent HIV protease inhibitor used to treat and prevent HIV/AIDS. Darunavir has a Ki of 1 nM for wild type HIV-1 protease.

Specifications

Chemical Name or Material Darunavir Ethanolate
CAS 635728-49-3
Color White
Purity Grade Notes Research
Molecular Formula C29H43N3O8S
Quantity 50 mg
Synonym TMC114 Ethanolate
Solubility Information DMSO : ≥ 50 mg/mL (84.21 mM)
SMILES O=C(O[C@@H]1[C@@]2([H])[C@@](OCC2)([H])OC1)N[C@@H](CC3=CC=CC=C3)[C@H](O)CN(S(=O)(C4=CC=C(N)C=C4)=O)CC(C)C.CCO
Molecular Weight (g/mol) 593.73
Formula Weight 593.73
Percent Purity 99.1%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.