missing translation for 'onlineSavingsMsg'
Learn More

Darunavir Ethanolate, MedChemExpress

Product Code. 16499797
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16499797 5 mg 5mg
16467288 10 mg 10mg
16433307 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16499797 Supplier Medchem Express Supplier No. HY17041
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Darunavir ethanolate (TMC114 Ethanolate) is a potent HIV protease inhibitor used to treat and prevent HIV/AIDS. Darunavir has a Ki of 1 nM for wild type HIV-1 protease.

Darunavir ethanolate (TMC114 Ethanolate) is a potent HIV protease inhibitor used to treat and prevent HIV/AIDS. Darunavir has a Ki of 1 nM for wild type HIV-1 protease.

Specifications

Chemical Name or Material Darunavir Ethanolate
CAS 635728-49-3
Color White
Purity Grade Notes Research
Molecular Formula C29H43N3O8S
Quantity 5 mg
Synonym TMC114 Ethanolate
Solubility Information DMSO : ≥ 50 mg/mL (84.21 mM)
SMILES O=C(O[C@@H]1[C@@]2([H])[C@@](OCC2)([H])OC1)N[C@@H](CC3=CC=CC=C3)[C@H](O)CN(S(=O)(C4=CC=C(N)C=C4)=O)CC(C)C.CCO
Molecular Weight (g/mol) 593.73
Formula Weight 593.73
Percent Purity 99.1%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.