missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DARS2 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-99371CL1
This item is not returnable.
View return policy
Description
DARS2 Polyclonal antibody specifically detects DARS2 in Human samples. It is validated for Western Blot
Specifications
| DARS2 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| CoraFluor 1 | |
| aspartate tRNA ligase 2, mitochondrial, Aspartate--tRNA ligase, aspartyl-tRNA synthetase 2, mitochondrial, aspartyl-tRNA synthetase, mitochondrial, AspRS, ASPRS. LBSL, EC 6.1.1, EC 6.1.1.12, FLJ10514, LBSL, MT-ASPRS, RP3-383J4.2 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human DARS2. | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 55157 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction