missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DARS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14989-1-AP-20UL
This item is not returnable.
View return policy
Description
Aspartyl-tRNA synthetase (DARS) is part of a multienzyme complex of aminoacyl-tRNA synthetases. Aspartyl-tRNA synthetase charges its cognate tRNA with aspartate during protein biosynthesis.Specifications
| DARS | |
| Polyclonal | |
| Unconjugated | |
| DARS | |
| Aspartate tRNA ligase, aspartyl tRNA synthetase, AspRS, DARS | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 116483, 1615, 226414 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P14868, P15178, Q922B2 | |
| Dars | |
| DARS Fusion Protein Ag6974 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction