missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DARS Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5143288
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognize reported isoform 1 (NP_803228.2) only. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.
Aspartyl-tRNA synthetase (DARS) is part of a multienzyme complex of aminoacyl-tRNA synthetases. Aspartyl-tRNA synthetase charges its cognate tRNA with aspartate during protein biosynthesis.
Specifications
| DARS | |
| Polyclonal | |
| Unconjugated | |
| DARS1 | |
| 5730439G15Rik; aspartate tRNA ligase 1, cytoplasmic; aspartate--tRNA ligase, cytoplasmic; Aspartyl-tRNA synthetase; aspartyl-tRNA synthetase, cytoplasmic; aspRS; Cell proliferation-inducing gene 40 protein; Dars; DRS1; HBSL; PIG40; testicular tissue protein Li 192 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 226414 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q922B2 | |
| DARS1 | |
| Peptide with sequence SRKGQEKPREIVD-C (aa 7-19). | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction