missing translation for 'onlineSavingsMsg'
Learn More

DARPP-32 Antibody (2A4G9) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-37302

 View more versions of this product

Product Code. 18182479

  • £435.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

DARPP-32 Monoclonal specifically detects DARPP-32 in Human samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DARPP-32
Monoclonal
1 mg/mL
Western Blot 1:500 - 1:2000, ELISA 1:10000
protein phosphatase 1, regulatory (inhibitor) subunit 1B, regulatory (inhibitor) subunit 1B (dopamine and cAMPregulated phosphoprotein, DARPP-32)
Mouse
23 kDa
0.1 mL
Breast Cancer, Neuronal Cell Markers, Neuroscience, Neurotransmission, Protein Phosphatase, Signal Transduction
84152
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Western Blot, ELISA
2A4G9
Unconjugated
PBS with 0.05% Sodium Azide
PPP1R1B
Purified recombinant fragment of human PPP1R1B (AA: 95-204) expressed in E. Coli.
Affinity Purified
RUO
Primary
Human
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only